Bottle, with Sealer Cap, LDPE, Dynalon
Supplier: Dynalon
605085-04 605085-06 605085-08
16354-806CS 169.47 CAD
16354-806 16354-808 16354-810
Bottle, with Sealer Cap, LDPE, Dynalon
Bottles Dropping Bottles
These low density polyethylene bottles are supplied with a sealer cap that snaps on easily after use and seals tightly.
Spout openings are standard 0.030", and can be enlarged by cutting back with scissors.