Laboratory Bottles, Wide Mouth, Polypropylene, Dynalon
Supplier: Dynalon
301625-0008 301625-0032 301625-0002
16354-836CS 823.02 CAD
16354-836 30620-190 30620-194
Laboratory Bottles, Wide Mouth, Polypropylene, Dynalon
Bottles Plastic Bottles
Leakproof, rigid, and translucent wide mouth polypropylene lab bottles can be autoclaved.
  • Excellent chemical resistance
  • Ideal for storage and sampling of liquids or solids
Delivery information: Supplied with a polypropylene closure.
Order Now
SPECIFICATIONS
Environmentally Preferable