Zipper Seal Bags, LDPE, Dynalon
Supplier: Dynalon
628425 628325 628335 628345 628365 628405 628415 628435
56766-314CS 454.93 CAD
56766-314 56766-300 56766-302 56766-304 30620-116 30620-118 30620-120 30620-122
Zipper Seal Bags, LDPE, Dynalon
Bags Sample Bags
LDPE zipper seal bags are ideal for collecting, storing, and protecting laboratory specimens and samples.
  • No printing on the bag allows for easier labeling and marking
  • Bags lock easily and securely with the zipper locking feature
Not suggested for use with liquids, bags provide many laboratory and industrial applications.