Polypropylene Carboys, Dynalon
Supplier: Dynalon
505634-2 505634-3 505634-1 505634-1 505634-2 505634-3
30620-246EA 226.75 CAD
30620-246 30620-248 30620-250 470200-406 470200-408 470202-528
Polypropylene Carboys, Dynalon
Bottles Carboys
Rounded octagonal shape for increased rigidity when full.
  • Heavy-duty autoclavable polypropylene construction
  • Offset neck enables easier filling and pouring
  • Comfortable blow molded handles allow easy carrying
Heavy-duty closures are compatible with other leading brands. Carboys feature a durable and leak-proof heavy-duty stopcock with boss fitting for extra strength. Stopcock features removable tube fitting for added convenience. Graduations are molded into the front of the product, allowing easy reading even when several carboys are used in a row.
Order Now
SPECIFICATIONS
Environmentally Preferable