Tweezers, PTFE, Dynalon
Supplier: Dynalon
316514-0002 316514-0001 316514-0000 316504-0001 316504-0002
24775-025CS 226.15 CAD
24775-025 24775-027 24775-029 24775-033 24775-035
Tweezers, PTFE, Dynalon
Forceps
Totally inert and non-contaminating forceps are manufactured from PTFE, making them suitable for use up to 260°C (500°F).