Hazardous Chemical Pourer, Dynalon
Supplier: Dynalon
310374-0002 310374-0003 310374-0004
13450-602EA 237.38 CAD
13450-602 13450-604 13450-606
Hazardous Chemical Pourer, Dynalon
Funnels Dropping Funnels
The hazardous and corrosive chemical pourer is manufactured fully from PTFE and features completely inert pouring surfaces
Even the neck-sealing ring is encapsulated in PTFE. The unique design allows direct and controlled pouring due to the incorporated breather vent. The pourer comes in four sizes to fit standard reagent bottles with neck sizes of GL45, 38, 32, and 30. A captured PTFE closure ensures contaminant free contents when not in use. Bottle not included.