Dipper, PTFE, Dynalon
Supplier: Dynalon
307024-0500
36495-114EA 753.66 CAD
36495-114
Dipper, PTFE, Dynalon
Samplers Sample Dippers
Totally inert and non-contaminating
Extendible screw-in shafts have a steel core for rigidity and are fully encapsulated in PTFE. The basic unit consists of a beaker with a single 24" (600mm) shaft.