3M™ XYZ-Axis Electrically Conductive Double Sided Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77808-12 77808-25 77808-63
102098-776EA 164.15 CAD
102098-776 102098-778 102098-780
3M™ XYZ-Axis Electrically Conductive Double Sided Tape, Electron Microscopy Sciences
Tapes
Conductive tape is an isotropically conductive pressure sensitive tape. The tape consists of a 3M™ adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance with very high conductivity. The Tape is an excellent choice when needing to bond your samples for all SEM work. Based upon four wire (Kelvin probe) resistance measurements made with crossed pieces of Foil/Type 9712 or 9713/Rigid plate construction using a 1.0×1.0" square piece of 3M™ tape Type 9712 or 9713. The rigid metal surface was prepared with a Scotch-More Product Information