Formvar Film on Gilder Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FF200F1-CU-TH FF200F1-CU-UL FF200F2-CU-UL FF200F2-CU-TH FF200F1-CU-ET FF200F2-CU-ET FF200F1-NI-UL FF200F2-NI-UL FF200F1-NI-TH FF200F2-NI-TH FF200F1-NI-ET FF200F2-NI-ET FF200F1-AU-TH FF200F2-AU-TH FF200F1-AU-ET FF200F2-AU-ET FF200F1-AU-UL FF200F2-AU-UL
103302-724EA 239.92 CAD
103302-724 103303-268 103303-270 103303-290 103303-372 103303-374 103303-550 103303-552 103303-826 103303-828 103303-884 103303-886 103305-304 103305-306 103305-388 103305-390 103305-926 103305-928
Formvar Film on Gilder Finder Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a Gilder Finder grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold slot Finder grids
Coated grids are optically checked and batch tested in the electron microscope.
Packaging: Packed in grid storage box.