Formvar Film on Slotted Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FF2010-CU-50 FF2010-NI-25 FF2010-NI-50 FF205-CU-25 FF205-CU-50 FF205-CU-UL FF2010-CU-UL FF2010-CU-TH FF2010-CU-ET FF2010-NI-UL FF2010-NI-TH FF2010-NI-ET FF2010-AU-UL FF205-AU-TH FF2010-AU-TH FF205-AU-ET FF2010-AU-ET
102100-816BX 274.16 CAD
102100-816 102100-818 102100-820 102100-826 102100-828 103303-462 103303-464 103303-584 103303-782 103303-784 103304-042 103304-254 103305-102 103305-208 103305-210 103305-300 103305-302
Formvar Film on Slotted Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a slotted grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold grids
Coated grids are optically checked and batch tested in the electron microscope.
Packaging: Packed in grid storage box.