Formvar Film on Single Hole Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FFGA400-CU-TH FFGA600-CU-TH FFGA800-CU-TH FFGA1000-CU-TH FFGA200-CU-UL FFGA600-CU-UL FFGA1000-CU-UL FFGA1500-CU-TH FFGA100-CU-ET FFGA600-CU-ET FFGA1500-CU-ET FFGA1500-NI-UL FFGA1500-NI-ET
103302-816EA 262.9 CAD
103302-816 103302-818 103302-820 103302-822 103303-472 103303-478 103303-482 103303-588 103303-790 103303-810 103303-822 103303-824 103304-274
Formvar Film on Single Hole Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a Single Hole grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper and nickel with 75, 100, 150, 200, 300, 400, 600, 800, 1000, or 1500 µm holes
  • Optically checked followed by batch testing in the EM
Coated grids are optically checked and batch tested in the electron microscope.
Packaging: Packed in grid storage box.