Spencer Stitch Scissors, Sklar
Supplier: Sklar
22-2835 22-2845 22-2834 22-2844
82030-138EA 170.77 CAD
82030-138 82030-140 89025-322 89025-328
Spencer Stitch Scissors, Sklar
Scissors
Sklar® Spencer Stitch Scissors are primarily used for suture removal.
These scissors have a small hook-shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.