SP Bel-Art Drying Tubes with Tube Fittings, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19960-0000 F19961-0000 F19962-0000 F19965-0000
20901-030PK 76.48 CAD
20901-030 20901-031 20901-032 47750-366
SP Bel-Art Drying Tubes with Tube Fittings, Bel-Art Products, a part of SP
Tubes Drying Tubes
Drying tubes with tube fittings can be filled with desiccant of choice.
  • Polyethylene construction
  • Virtually unbreakable
Each unbreakable polyethylene straight tubes, 16 mm int.Ø. and 19 mm ext.Ø. (⅝×¾"), is supplied with two polypropylene fittings with serrated tapered end that accept 6.4 to 9.5 mm (¼ to ⅜") int.Ø. tubing. Drying agent not included.