API Precision Combined Form Hydrometer, Thermco
Supplier: Thermco
GW6000S GW6001S GW6001XS GW6002S GW6003S GW6004S GW6005S GW6006S
10053-570EA 145.23 CAD
10053-570 10053-572 10053-574 10053-576 10053-578 10053-580 10053-582 10053-584 10053-586 10053-588 10053-590 10053-592 10053-594
API Precision Combined Form Hydrometer, Thermco
Hydrometers
Precison API combined form hydrometers made to meet API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and mounted in the body of the hydrometer. Length of the hydrometer is 15 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.