API Precision Combined Short Form Hydrometer, Thermco
Supplier: Thermco
GW2500CS GW2501CS GW2502CS GW2503CS GW2504CS GW2505CS GW2507CS GW2508CS GW2509CS
10053-300EA 129.01 CAD
10053-300 10053-302 10053-304 10053-306 10053-558 10053-560 10053-564 10053-566 10053-568
API Precision Combined Short Form Hydrometer, Thermco
Hydrometers
Precison API combined form hydrometers made to meet API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 20 to 120 degrees farenheit.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
SPECIFICATIONS
Environmentally Preferable