API Precision Short Plain Form Hydrometer, Thermco
Supplier: Thermco
GW2500 GW2501 GW2502 GW2503 GW2504 GW2505 GW2506 GW2507 GW2508 GW2509
10053-280EA 74.39 CAD
10053-280 10053-282 10053-284 10053-286 10053-288 10053-290 10053-292 10053-294 10053-296 10053-298
API Precision Short Plain Form Hydrometer, Thermco
Hydrometers
Precison API plain form hydrometers made to meet API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers.
Length of the hydrometer is 6 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.