ASTM/API Precision Combined Form +30/180 ºF Hydrometer, Thermco
Supplier: Thermco
GW041HMS GW042HMS GW043HMS GW044HMS GW045HMS GW051HMS GW052HMS GW053HMS GW054HMS GW055HMS GW056HMS GW057HMS GW058HMS
10053-276EA 134.25 CAD
10053-276 10053-278 10053-532 10053-534 10053-536 10053-538 10053-540 10053-542 10053-544 10053-546 10053-548 10053-550 10053-552 10053-554 10053-556
ASTM/API Precision Combined Form +30/180 ºF Hydrometer, Thermco
Hydrometers
Precison ASTM-like combined form hydrometers made to meet ASTM and API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 30 to 180 degrees farenheit and is mounted in the body of the hydrometer. Length of hydrometer is 380mm.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.