ASTM/API Precision Combined Form 0/150 ºF Hydrometer, Thermco
Supplier: Thermco
GW041HLS GW042HLS GW043HLS GW044HLS GW045HLS GW051HLS GW052HLS GW053HLS GW054HLS GW055HLS GW056HLS GW057HLS GW058HLS GW059HLS
10053-502EA 134.3 CAD
10053-502 10053-504 10053-506 10053-508 10053-510 10053-512 10053-514 10053-516 10053-518 10053-520 10053-522 10053-524 10053-526 10053-528 10053-530
ASTM/API Precision Combined Form 0/150 ºF Hydrometer, Thermco
Hydrometers
Precison ASTM-like combined form hydrometers made to meet ASTM and API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and is mounted in the body of the hydrometer. Length of hydrometer is 380mm.
Used to measure the density according to API degree scale for petroleumMore Product Information