ASTM/API Precision Plain Form Hydrometer, Thermco
Supplier: Thermco
GW003H GW004H GW005H GW006H GW007H GW008H GW009H GW010H GW021H GW022H GW023H GW024H GW025H GW026H GW028H GW029H GW030H GW031H GW001H GW002H GW032H GW033H GW034H GW036H GW037H GW040H
10053-150EA 79.77 CAD
10053-150 10053-152 10053-154 10053-156 10053-158 10053-160 10053-162 10053-164 10053-166 10053-168 10053-170 10053-172 10053-174 10053-176 10053-180 10053-182 10053-184 10053-186 10053-272 10053-274 10053-484 10053-486 10053-488 10053-490 10053-492 10053-494 10053-496 10053-498 10053-500
ASTM/API Precision Plain Form Hydrometer, Thermco
Hydrometers
Precison ASTM plain form hydrometers made to meet ASTM and API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 0.1 degrees.
Tall form, 330mm, have 12 degree ranges; Short form, 163mm, have 6 degree ranges.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.