Ultra Hygiene Squeegees, 16", Remco Products
Supplier: Remco Products
71401 71402 71403 71404 71405 71406 71407 71408 71409
76008-076EA 50.04 CAD
76008-076 76008-078 76008-080 76008-082 76008-084 76008-086 76008-088 76008-090 76008-092
Ultra Hygiene Squeegees, 16", Remco Products
Mops
The ultra hygiene squeegee is particularly suited for ”sweeping” smooth, wet floors to remove large amounts of dirt, as the single squeegee blade design is extremely easy to clean and sanitize.
This squeegee model is suitable for use in food processing facilities, large kitchens, hospitals and other areas where hygiene is critical. Polypropylene block material. Rubber blade material.
Certifications: FDA compliant raw material.