GOLD BRAND API High Precision Combined Form Hydrometer, Thermco
Supplier: Thermco
ACC6001S ACC6001XXS ACC6003S ACC6004S ACC6005S ACC6006S ACC6007S
10053-134EA 181.12 CAD
10053-134 10053-136 10053-138 10053-140 10053-142 10053-144 10053-146 10053-148 10053-268 10053-270
GOLD BRAND API High Precision Combined Form Hydrometer, Thermco
Hydrometers
High precison API combined form hydrometers made to exceed API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees farenheit. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and is mounted in thebody of the hydrometer. Length of hydrometer is 15 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.