PB-900™ Programmable Jar Tester, Phipps & Bird
Supplier: Phipps & Bird
7790-900 7790-910 7790-950 7790-960 7790-980
89496-308EA 8843.69 CAD
89496-308 89496-310 89496-312 89496-314 89496-316
PB-900™ Programmable Jar Tester, Phipps & Bird
Shakers Vortex Mixers
The Phipps & Bird Jar Tester has been re-engineered to improve its performance and bring it into compliance with CE and cTUVus requirements
The motor has been improved to a stepper motor drive system. Additional features include LED lighting modules and complete 24VDC with included adapter that will power the machine from 100 to 240V sources.