Vikan® High-Low Brushes / Scrubbing Brooms, Remco Products
Supplier: Remco
Vikan®
70474 70472 70476 70475 70473
76008-004EA 66.83 CAD
76008-004 76008-006 76008-008 76007-966 76008-010
Vikan® High-Low Brushes / Scrubbing Brooms, Remco Products
Brooms
The angled shape of this scrubbing broom was designed to clean the area where the wall meets the floor, but also effective for cleaning the floor itself.
  • Block material is polypropylene; bristle material is polyester
  • Dishwasher safe
  • Autoclavable up to 250°F
Its bristles are gentle enough to be used on equipment. It is a popular choice in meat processing facilities, canneries, mills, wineries and breweries—and generally any moist environment that creates difficult cleaning challenges
Certifications: FDA compliant raw material.