Nunc® Externally-Threaded Universal Tubes, 1.8 ml, Thermo Scientific
Supplier: Thermo Fisher Scientific
374501 374502 374503 331830
89233-710CS 1040.79 CAD
89233-710 89233-712 89233-714 89233-716
Nunc® Externally-Threaded Universal Tubes, 1.8 ml, Thermo Scientific
Tubes Test Tubes
Versatile Nunc® 1.8 ml externally-threaded universal tubes can be used inside a large storage system, or in smaller scale applications such as traditional cryostorage in dewars.
All tubes and trays are supplied free from DNA, RNAse, DNAse, and endotoxins.
Certifications: IATA certification ensures that shipped samples will not leak.
Packaging: Available racked or in bulk, with 2D barcodes or non-coded.