Two-Speed Lab Blenders, Waring
Supplier: Waring
7011S 7011G 7011G
74790-002EA 1423.2 CAD
74790-002 58977-103 58977-089
Two-Speed Lab Blenders, Waring
Homogenizers Standard Laboratory Blenders
This versatile laboratory blender is applicable for most lab work, providing no-load speeds of 18,000rpm and 22,000rpm
Available with either a heat-resistant glass, or stainless steel 1L container. Both have a handle and two-piece vinyl and styrene lid. Motor housing is epoxy-coated. Rated Amps: 3.6; 120V, 50/60Hz; 0.40HP. UL listed. Also available in heavy-duty versions.