Vikan® Waterfed Deck Scrubs, Stiff, Remco Products
Supplier: Remco Products
70416 70412 70413 70414 70415
76007-612EA 51.24 CAD
76007-612 76007-952 76007-954 76007-956 76007-958
Vikan® Waterfed Deck Scrubs, Stiff, Remco Products
Brooms
Extremely stiff, short-bristled scrubbing broom.
  • Block material is polypropylene; bristle material is polyester
  • Small size makes it ideal for cleaning in narrow spaces
  • Max. temperature: 250°F
When dirt is stubborn and sticky, this broom can withstand heavy downward force from the user.
Head depth: 3"
Certifications: FDA compliant raw material.