Vikan® Tank Brushes, Medium, Remco Products
Supplier: Remco Products
70392 70393 70394 70395 70396
76007-942EA 63.11 CAD
76007-942 76007-944 76007-946 76007-948 76007-950
Vikan® Tank Brushes, Medium, Remco Products
Brushes
Ideal for cleaning tanks and vats, and is also suitable for cleaning drain gulleys.
  • Block material is polypropylene; bristle material is polypropylene
  • Dishwasher safe
  • Autoclavable up to 250 °F
May be used with any European-thread handle from Vikan.
Certifications: FDA compliant raw material.
Order Now
SPECIFICATIONS
Environmentally Preferable