GOLD BRAND ASTM/API High Precision Plain Form Hydrometer, Thermco
Supplier: Thermco
ACC001H ACC002H ACC003H ACC004H ACC005H ACC006H ACC007H ACC008H ACC009H ACC010H ACC021H ACC022H ACC023H ACC025H ACC026H ACC027H ACC028H ACC029H ACC030H ACC031H ACC033H ACC039H
10053-046EA 107.66 CAD
10053-046 10053-048 10053-050 10053-052 10053-054 10053-056 10053-058 10053-060 10053-062 10053-064 10053-066 10053-068 10053-070 10053-072 10053-074 10053-076 10053-078 10053-080 10053-082 10053-084 10053-086 10053-088 10053-090 10053-092 10053-094 10053-096 10053-098 10053-100 10053-102 10053-104
GOLD BRAND ASTM/API High Precision Plain Form Hydrometer, Thermco
Hydrometers
High precison ASTM plain form hydrometers made to exceed ASTM and API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 0.1 degree.
Tall form, 330 mm, has 12 degree ranges; Short form, 163 mm, has 6 degree ranges.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Ordering information: Hard square plastic tube included.