ASTM-Approved Non-Mercury Tank Gauging Thermometers, Thermco
Supplier: Thermco
ACC272WB3 ACC273WB1S ACC273WB2 ACC273WB3 ACC272WB1 ACC272WB2
10053-036EA 193.53 CAD
10053-036 10053-038 10053-040 10053-042 10053-480 10053-482
ASTM-Approved Non-Mercury Tank Gauging Thermometers, Thermco
Thermometers Analog Thermometers
Wood Brass Cup model, case made from a hardwood that prevents absorbtion of the petroleum sample
Thermometers supplied with each case are ASTM approved, non-mercury blue spirit; remounted on spring clips and can be replaced if breakage occurs. Three ranges available in both celcius and farenheit.
The attached brass cup is non-sparking and meets ASTM and API requirements.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Woodback Cup Case Thermometer Only. Case is available separately.