ASTM Approved Non-Mercury 'Precision' Thermometers, Thermco
Supplier: Thermco
ACC065FBLS ACC062CBLS ACC062FBLS ACC063CBLS ACC063FBLS ACC066CBLS ACC066FBLS ACC067CBLS ACC067FBLS ACC064CBLS ACC064FBLS ACC065CBLS
10052-980EA 578.06 CAD
10052-980 10052-982 10052-984 10052-986 10052-988 10053-220 10053-222 10053-224 10053-226 10053-454 10053-456 10053-458
ASTM Approved Non-Mercury 'Precision' Thermometers, Thermco
Thermometers Analog Thermometers
ASTM approved non-mercury blue spirit "precision" thermometers
Tenth of a scale division. The ice piont enables the user to validate and update the calibration document. Rages covered are from -38 to 155 degrees C and -36 to 311 degrees F.
Each incorporates an ice point on the scale, calibrated against NIST standards and certified at one temperature point.
Certifications: ASTM approved/ISO17025/ILAC-MRA accredited lab.
Ordering information: Accessories not included.