SP Bel-Art Secador® 1.0, 2.0, 3.0, 4.0 Gas-Purge Desiccator Cabinets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42072-1002 F42073-1002
47750-996EA 29883.44 CAD
47750-996 47750-998
SP Bel-Art Secador® 1.0, 2.0, 3.0, 4.0 Gas-Purge Desiccator Cabinets, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture- and UV-sensitive products.
  • Sturdy construction
  • Easy and reliable storage
  • Protects moisture- and UV-sensitive products
  • Polycarbonate construction
Polycarbonate construction features a UV light blocker and is resistant to staining, crazing, and chemical attack.
Rear-mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D. Large doors maximize access to interior space and have a patented seal design, tabMore Product Information