Wet-Strengthened Qualitative Filter Paper, Grade 909, Ahlstrom
Supplier: Ahlstrom
9090-1100 9090-1250 9090-1500 9090-1850 9090-2400
47749-722PK 31.3 CAD
47749-722 47749-724 47749-726 47749-728 47749-730
Wet-Strengthened Qualitative Filter Paper, Grade 909, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 909 has a rough surface, a medium flow rate, and is a tighter version of Grade 962.