Black Qualitative Filter Paper, Grade 8613, Ahlstrom
Supplier: Ahlstrom
8613-1250 8613-0900 8613-0425 8613-0470 8613-0471 8613-0240 8613-0550 8613-1500 8613-1850 8613-2050
28320-555PK 205.04 CAD
28320-555 28342-010 28342-012 28342-020 28342-052 47745-870 47745-872 47745-876 47745-878 47746-930
Black Qualitative Filter Paper, Grade 8613, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom 8613 has a smooth black finish designed for use in applications requiring visual examination of particles.