Cole-Parmer® WBU-200 Series Liquid Calibration Baths, Antylia Scientific
Supplier: Antyila Scientific
16104-50 16105-04
14228-908EA 3533.07 CAD
14228-908 14228-914
Cole-Parmer® WBU-200 Series Liquid Calibration Baths, Antylia Scientific
Circulating Thermostats Open Circulator Systems Calibration Circulators
Compact and accurate water baths for external circulation or temperature calibration.
  • Fully insulated baths offer excellent heat retention
  • Temperature stability of ±0.005 °C depending on choice of control unit
These compact calibration baths offer excellent stability over the entire temperature range. Use these baths for external circulation to maintain temperatures of samples in viscometers, photometers, refractometers, fermenters and other reaction vessels.
All models offer high pump performance and exceptional thermal stability from −35 to 250 °C. Add a flow cooler to the system to achieve temperatures down to −35 °C. The water baths are fully insulated on all sides and base and are fitted with a cooling coil for connection to a cold water supply for use at temperatures around ambient.
Ordering information: An immersion circulator is required for liquid calibration baths to operate—order Cole-Parmer IC-300 series or other compatible immersion circulators separately. Supplied with lid, drain tap, carry handles, a cooling coil (with bung) and hole to position a certified sensor.