Wet-Strengthened Qualitative Filter Paper, Grade 961, Ahlstrom
Supplier: Ahlstrom
9610-0550 9610-0700 9610-0900 9610-1100 9610-1250 9610-1500 9610-1850 9610-2050 9610-2400 9610-3200 9610-3850 9610-4000 9610-4500 9610-5000 9618-4657 9611-2020
47749-762PK 24.85 CAD
47749-762 47749-764 47749-766 47749-768 47749-770 47749-772 47749-774 47749-776 47749-778 47749-780 47749-782 47749-784 47749-786 47749-788 47749-856 28342-024
Wet-Strengthened Qualitative Filter Paper, Grade 961, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom 961 is a fast filtering grade with a creped surface and extremely high loading capacity.
It is recommended for the filtration of coarse and gelatinous precipitates.