Pleated (Fluted) Filter Paper Grade 562, Ahlstrom
Supplier: Ahlstrom
5620-1250 5620-1500 5620-1850 5620-2400 5620-3200 5620-3850
47745-852PK 263.23 CAD
47745-852 47745-854 47745-856 47745-858 47745-860 47745-862
Pleated (Fluted) Filter Paper Grade 562, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 562 is the pre-pleated version of Grade 962.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.