Pleated (Fluted) Filter Paper, Grade 513, Ahlstrom
Supplier: Ahlstrom
5130-1250 5130-1500 5130-2400 51303200
47745-840PK 228.17 CAD
47745-840 47745-842 47745-844 71440-004
Pleated (Fluted) Filter Paper, Grade 513, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 513 is the pre-pleated version of Grade 613.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.