Glass Microfiber Filter Paper, Grade 193, Ahlstrom
Supplier: Ahlstrom
1930-0470 1930-0900 1930-1100 1930-1250 1930-1420 1930-1500
12000-108PK 243.57 CAD
12000-108 12000-110 12000-112 12000-114 12000-116 12000-118
Glass Microfiber Filter Paper, Grade 193, Ahlstrom
Filters Glass Fiber Filters
Grade 193 is a binder-free microfiber glass filter made with acid-treated borosilicated glass microfibers with inherently low levels of barium and zind.
The filter media contains no binders or other additives that may cause interference with samples. It is recommended for use in the Toxicity Characteristic Leaching Procedure (TCLP) as described in EPA Method 1311.