Hardened Ashless Quantitative Paper, Grade 75, Ahlstrom
Supplier: Ahlstrom
0750-0900 0750-1100 0750-1250 0750-1500 0750-0210 0750-0240 0750-0425 0750-0550 0750-0700 0750-1850 0750-2400 0750-2700 0750-3200 0750-5000
47745-746PK 101.77 CAD
47745-746 47745-748 47745-750 47745-752 47749-478 47749-480 47749-482 47749-484 47749-486 47749-488 47749-490 47749-492 47749-494 47749-496
Hardened Ashless Quantitative Paper, Grade 75, Ahlstrom
Filters Filter Papers Quantitative Filter Paper
Ahlstrom Grade 75 is a medium flow rate hardened ashless (Ash 0.006%) filter paper suitable for quantitative gravimetric analysis in analytical techniques requiring increased chemical resistance, wet strength and handling capacity.