Hardened Ashless Quantitative Paper, Grade 95, Ahlstrom
Supplier: Ahlstrom
0950-0550 0950-0700 0950-0900 0950-1100 0950-1250 0950-1500 0950-0210 0950-0240 0950-0425 0950-1850 0950-2400 0950-2700 0950-3200 0950-4000 0950-5000
47745-760PK 70.89 CAD
47745-760 47745-762 47745-764 47745-766 47745-768 47745-770 47749-508 47749-510 47749-512 47749-514 47749-516 47749-518 47749-520 47749-522 47749-524
Hardened Ashless Quantitative Paper, Grade 95, Ahlstrom
Filters Filter Papers Quantitative Filter Paper
Ahlstrom Grade 95 is an extremely fine retention, hardened ashless (Ash 0.006%) filter paper suitable for quantitative gravimetric analysis in analytical techniques requiring increased chemical resistance, wet strength and handling capacity.