BX™ series protective eyewear features a soft fully adjustable nose bridge, lens angle adjustment, and adjustable length temples for extended wear.
- Nose bridge is cushioned for softness and comfort
- Sleek, contoured design offers an attractive appearance along with functional eye protection
- Impact-resistant polycarbonate lenses absorb 99.9% UV rays
The ability to customize the safety glasses to the wearer makes them more pleasant to use over extended time periods.
Eye protection is important to safeguard the user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. All 3M™ protective eyewear combines safety features with comfort enhancements.
These safety glasses are recommended for use in assembly and mechanical, chipping, chiseling, cleaning, drilling, grinding, machining, masonry, metal repair, painting, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals, and transportation.
Certifications: ANSI Z87.1-2003. CSA Z94.3-2007.