Vikan® Aluminium Handles, 51", Remco Products
Supplier: Remco Products
29357 29358 29359 29352 29353 29354 29355 29356
76006-612EA 47.34 CAD
76006-612 76006-614 76006-616 76007-210 76007-212 76007-214 76007-216 76007-218 76007-220
Vikan® Aluminium Handles, 51", Remco Products
Brooms
This standard broom handle can be used with all brooms, squeegees and scrapers.
  • Aluminum and polypropylene materials
The handle with the attached item should reach to the chin of the user to prevent occupational back injuries.
Certifications: FDA compliant raw material.