CPEP History and Fate of the Universe Charts
Supplier: CONTEMPORARY PHYSICS EDUCATION
528441 528400 528412 528431
470218-780EA 21.8 CAD
470218-780 470218-782 470218-784 470218-786
CPEP History and Fate of the Universe Charts
Charts
This colorful, graphically rich chart illustrates and summarizes what is now known about the history and fate of the universe.
Developed by scientists at the Lawrence Berkeley National Laboratory (Berkeley Lab) in collaboration with the Contemporary Physics Education Project (CPEP) and George Smoot, Nobel prize winner for 2006, the chart is crammed with information covering a broad range of cosmological topics.
The centerpiece is an evolutionary timeline that takes viewers from 10-44 seconds, when the universe was much smaller than a proton, to the current era, about 14 billion years later,More Product Information