CPEP Nuclear Science Charts
Supplier: CONTEMPORARY PHYSICS EDUCATION
296231 296200 296241 296212
470073-046EA 49.35 CAD
470073-046 470072-810 470122-542 470076-274
CPEP Nuclear Science Charts
Charts
Visualize nuclear science.
  • Describes basic nuclear processes and the modern picture of the nucleus and then applies them to the formation of the Universe
Nuclear Science, part of the highly popular Contemporary Physics Education Project (CPEP) series, explores the structure, properties and interactions of atomic nuclei.
It illustrates the known isotopes with a chart of the nuclides in graphic form, traces radioactive decay from alpha to gamma and shows how new elements are found. Many of the applications of nuclear science are described.