Buchner Filter Funnels, Extra Capacity, Chemglass
Supplier: Chemglass
CG-1402-L-01 CG-1402-L-02 CG-1402-L-06 CG-1402-L-03 CG-1402-L-04 CG-1402-L-05 CG-1402-L-07 CG-1402-L-08
89028-454EA 185.05 CAD
89028-454 89028-456 89028-464 89028-458 89028-460 89028-462 89028-466 89028-468
Buchner Filter Funnels, Extra Capacity, Chemglass
Funnels Filtration Funnels Büchner Funnels
With an increased height above the disc by 33% for greater capacity
Not recommended for use where pressure would exceed 15 PSI.