Plastic Coated Solvent Delivery Bottles, Chemglass
Supplier: Chemglass
CG-1125-02 CG-1125-01
80066-374EA 485.93 CAD
80066-374 80066-372
Plastic Coated Solvent Delivery Bottles, Chemglass
Bottles Solution Bottles
Solvent reservoir for use in HPLC mobile phase delivery.
Outer joint at the top on all sizes is polished, thereby making an excellent seal between the PTFE cap and the bottle. Optional stopper permits convenient storage of solvent.