DiamondⓇ White Glass Microscope Slides, Plain, Globe Scientific
Supplier: Globe Scientific
1380-10 1384-10
10118-612CS 1363.8 CAD
10118-612 10118-618
DiamondⓇ White Glass Microscope Slides, Plain, Globe Scientific
Slides Microscope Slides
Globe scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
  • Produced from the finest quality optical white glass
  • Dimensions: 25×75 mm
  • Thickness: 1 mm
DiamondⓇ White Glass plain microscope slides are available with 90° or clipped corners. The slides are pre-cleaned and supplied ready for use.
Packaging: Slides are packaged 72 per box, 20 boxes per case (10 gross).