Large Scoops, Remco Products
Supplier: Remco Products
65001 65003 65004 65005 65006 65007 65008
76007-790EA 16.4 CAD
76007-790 76007-794 76007-796 76007-798 76007-800 76007-802 76007-804
Large Scoops, Remco Products
Scoops
Fully color-coded large polypropylene scoops are great for handling bulk materials
The solid construction makes them extremely durable. Large scoops have no seams or cracks to help prevent bacterial growth.
Certifications: FDA Compliant Raw Material.