Conductivity Cells, IDS, for Multi 3410 / 3420 / 3430, WTW
Supplier: WTW
301710 301711 301720
97042-952EA 2203.66 CAD
97042-952 97042-954 97042-960
Conductivity Cells, IDS, for Multi 3410 / 3420 / 3430, WTW
Electrodes Conductivity Sensors
High quality, enhanced sensor technology combined with most modern measurement electronics.
  • IDS cells include their serial number and their calibration records
  • Two different cells covering all applications from pure water to highly conducting solutions
  • Temperature Range: 0 to +100 °C
Various cable lengths available.