Oxygen Meters, Handheld, Oxi 3205 / 3210 / 3310, WTW
Supplier: WTW
2BA100 2BA101 2BA300 2BA301
97043-118EA 3049.31 CAD
97043-118 97043-120 97043-126 97043-130 97043-134 97043-136
Oxygen Meters, Handheld, Oxi 3205 / 3210 / 3310, WTW
Electrochemistry Meters Oxygen Meters
Waterproof handheld meters designed for unlimited outside use.
  • Large display allowing excellent readability, even under bad lighting conditions
  • Robust housing and sealed silicone keypad with protection class IP 66/67
  • Up to 2500 hour continuous operation time with one battery set
For challenging measurements in the field and plant, e.g
The Oxi 3205 is a small version of the Oxi 3210 without memory but including simplified functions.
For professional field measurements requiring documentation and data transfer. The Oxi 3310 IDS models enable digital measuring of optical DO. The optical DO sensor FDO® 925 is not only recommended for BOD measuring, but also for field and process applications.
Same specifications as Oxi 3210, with:
Extended memory for 5000 GLP-compliant data
Time-controlled datalogger with adjustable intervals for long-term monitoring
Waterproof mini USB interface to a PC.
Ordering information: All oxygen meters are available in sets including sensors and accessories.